Learn More
Cayman Chemical SphngosInd16 1 10mg
SDP
Supplier:  Cayman Chemical 2235610 MG
Encompass_Preferred
An atypical sphingolipid; inhibits PKC in a mixed micelle activity assay; reduces PMA-induced superoxide production in isolated human neutrophils and reduces the growth of CHO cells (IC50s = 1 and 3.2 µM, respectively); plasma levels of sphingolipids containing sphingosine (d16:1) are associated with dietary intake of saturated fatty acids and protein in ethnic Chinese individuals; plasma levels of sphingolipids containing sphingosine (d16:1) are decreased in patients with type 2 diabetes
Catalog No. 50-296-4320