Learn More
Medchemexpress LLC Sphondin | 483-66-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24295MG
Encompass_Preferred
Sphondin is a compound that demonstrates an inhibitory effect on the increase of COX-2 protein levels and PGE2 release induced by IL-1β in A549 cells. It is intended for research use only and not sold to patients. It is an off-white to light yellow solid with a molecular weight of 216.19 and a purity of 99.70%.
  • Inhibits COX-2 protein expression and PGE2 release
  • Off-white to light yellow solid appearance
  • High purity of 99.70%
  • Suitable for research applications
Catalog No. 50-004-20748