Learn More
Medchemexpress LLC Sphondin | 483-66-9 | MFCD00597230 | 99.7% | 216.19 g/mol | C12H8O4 | 10 MG

Supplier: Medchemexpress LLC HYN242910MG
Sphondin is a natural furanocoumarin research reagent that inhibits IL-1β-induced COX-2 protein upregulation and PGE2 release in A549 cells. Provided as a high-purity solid (C12H8O4; MW 216.19 g/mol) for in vitro research; refer to supplier documentation for storage and handling.
- Inhibits IL-1β-induced COX-2 protein increase and PGE2 release in A549 cells.
- High purity suitable for biochemical and cell-based assays.
- Molecular formula C12H8O4; molecular weight 216.19 g/mol.
- Typical physical form: solid; soluble in DMSO.
- Recommended storage: sealed, away from moisture and light; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month).
- Available in a 10 mg pack size for small-scale experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.