Learn More
MSC SPI ZZ Plus Plug Gage, 0.9480 in. Diameter, Black Oxide, 1 ea.
Supplier:  MSC 56243041
Encompass
This Class ZZ Plus plug gage provides high-precision measurement for industrial quality control applications. It is engineered for go inspection, setting micrometers, and calibrating verniers and other measuring devices in accordance with ASME B89.1 standards.
  • Diameter of 0.9480 in.
  • Tolerance of +0.000200 in.
  • Durable black oxide coating
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-14481