Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5940 in. Diameter, Black Oxide, 1 ea
Supplier:  MSC 56239460
Encompass
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. Featuring a nominal diameter of 0.5940 in., it provides a reliable standard for go/no go testing in various industrial and metalworking environments.
  • Nominal diameter of 0.5940 in.
  • Tolerance of +0.000200 in.
  • Black oxide coating for enhanced durability
Catalog No. 50-005-11982