Learn More
MSC SPI Swiss Precision Instruments Class ZZ Plug Gage, Black Oxide, 0.179 in. Diameter, -0.0002 in. Tolerance, 1 ea.
Supplier:  MSC 56245285
Encompass
This SPI Class ZZ plug and pin gage is designed for precise measurement and inspection applications. Featuring a black oxide finish for enhanced durability and corrosion resistance, it provides reliable performance in a variety of industrial settings.
  • Diameter of 0.179 in.
  • Tolerance of -0.000200 in.
  • Constructed with a black oxide finish
  • Class ZZ precision rating
Catalog No. 50-005-11466