Learn More
MSC SPI Plug Gage, 0.832 in. Diameter, Black Oxide, 1 ea.
Supplier:  MSC 56251820
Encompass
This Class ZZ plug and pin gage is designed for precise measurement applications, featuring a diameter of 0.832 in. and a length of 2 in. Constructed from hardened drill rod, this tool is built for durability and consistent performance in metalworking and inspection tasks.
  • Hardened drill rod construction with a hardness of RC 60 to 62
  • Black oxide finish provides corrosion resistance and allows for visual monitoring of gage wear
  • Tolerance of 0.0002 in.
  • Made in the USA
Catalog No. 50-005-10675