Learn More
MSC SPI Swiss Precision Instruments Plug Gage, 0.24 in. Diameter, 0.0002 in. Tolerance, Black Oxide Finish, 1 ea.
Supplier:  MSC 84500792
Encompass
This Class ZZ Plus plug gage is designed for precise diameter measurements, featuring a 0.2400 in. diameter and a tolerance of 0.0002 in.
  • Provides a 0.2400 in. nominal diameter for accurate inspection tasks.
  • Constructed with a black oxide finish for enhanced durability.
  • Maintains a tight tolerance of 0.0002 in.
Catalog No. 50-005-15218