Learn More
MSC SPI Pin Gage, Black Oxide Finish, 0.865 in. Diameter, 1 ea.
Supplier:  MSC 56242209
Encompass
This black oxide pin gage is designed for precision measurements, offering a diameter of 0.8650 in. with a tolerance of +0.000200 in. Its construction ensures reliability in various metrology applications.
  • Black oxide coating prevents corrosion and indicates gage wear
  • Manufactured for high precision
  • Suitable for measuring applications
Catalog No. 50-005-15055