Learn More
MSC Wera Soft Face Hammer, 33 mm, Semi-hard Urethane, Ashwood Handle, 1 ea.
Supplier:  MSC 84500206
Encompass
The 102 series soft-faced hammer features a semi-hard urethane head designed to provide powerful impact while protecting delicate work surfaces from damage. This tool features a high-quality ashwood handle, ensuring durability and comfort during extended use. The design incorporates an inseparable, secure connection between the hammer head and handle, making it suitable for demanding applications in automotive, engineering, and car body work.
  • Semi-hard urethane head for surface protection
  • High-quality ashwood handle for durability
  • Inseparable and secure connection between handle and head
  • Ideal for sharp-edged workpieces
  • Suitable for automotive, engineering, and car body applications
  • Low spark model
  • Antimagnetic construction
Catalog No. 50-005-15001