Learn More
MSC SPI Class ZZ Minus Pin Gage, 0.4470 in. Dia, Black Oxide Coating, 1 ea.
Supplier: MSC 56247968
This Class ZZ plug and pin gage is designed for precise measurement and inspection applications. It features a hardened steel construction for durability and long-lasting accuracy in industrial settings.
- Diameter of 0.4470 in.
- Tolerance of 0.000200 in.
- Black oxide coating
- Hardened to RC 60-62
- Length of 2 in.