Learn More
MSC SPI Swiss Precision Instruments Class ZZ Minus Pin Gage, 0.661 in. Diameter, Black Oxide, 1 ea
Supplier:  MSC 56250103
Encompass
This SPI Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, setting micrometers, and acting as a Go/No-Go inspection tool for various precision measuring devices. Constructed from hardened drill rod, this gage offers a durable and reliable solution for rigorous shop floor applications.
  • Diameter of 0.661 in.
  • Tolerance of 0.000200 in.
  • Hardened drill rod construction with RC 60-62 hardness
  • Black oxide coating for corrosion resistance and wear indication
  • Length of 2 in.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers and use in No-Go inspection
Catalog No. 50-005-14891