Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7060 in. Diameter, Black Oxide, 1 ea.
Supplier:  MSC 56240591
Encompass
This Class ZZ Plus plug gage is designed for precise hole diameter verification and inspection tasks. Featuring a black oxide finish, it offers enhanced wear resistance and durability in industrial measuring applications.
  • Diameter of 0.7060 in.
  • Tolerance of +0.000200 in.
  • Black oxide coating for improved surface protection
Catalog No. 50-005-14482