Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.7270 in, Black Oxide
Supplier:  MSC 56250764
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.7270 in diameter and a durable black oxide finish, ensuring reliability and accuracy in various industrial settings.
  • Designed for precise gaging.
  • Features a 0.7270 in diameter.
  • Equipped with a black oxide finish.
  • Maintains a tight tolerance of 0.000200 "
Catalog No. 50-005-31823