Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.7490 in, Black Oxide
Supplier:  MSC 56250988
Encompass
This Class ZZ Minus Plug & Pin Gage is engineered for precise measurement and inspection tasks. Featuring a diameter of 0.7490 inches and a durable black oxide finish, it provides reliable performance in various applications.
  • Engineered for precise measurement and inspection tasks.
  • Diameter of 0.7490 in.
  • Tolerance of 0.000200 in.
  • Durable black oxide coating.
  • Manufactured for consistent quality.
Catalog No. 50-005-31779