Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.1180 in Dia, 0.000200 in Tolerance, Steel
Supplier:  MSC 78383031
Encompass
This plug gage is designed for precise measurement applications, offering a high level of accuracy and durability. It features a specific diameter and tolerance, making it suitable for quality control and inspection tasks.
  • Made from durable steel material.
  • Measures 0.1180 inch in diameter.
  • Features a tight tolerance of 0.000200 "
  • Ideal for accurate Go/No-Go measurement.
Catalog No. 50-005-31684