Learn More
MSC SPI Dial Height Gage, 12 in, Stainless Steel, 0.001 in Graduation, 0.0015 in Accuracy
Supplier: MSC 69549186
This dial height gage features a 12 inch measuring capacity with a 0.001 inch graduation and an accuracy of 0.0015 " Constructed from stainless steel, it includes a dial and counter display for accurate and easy-to-read fine and coarse measurements. The resettable digital counter provides individual displays for upward and downward measurements from a preset zero, and the gage comes with NIST traceability certification.
- 0.001 in graduation
- Accurate to 0.0015 in
- Dial and counter display for accurate and easy-to-read measurements
- NIST traceability certification
- Fine adjustment capability
- Constructed from stainless steel
- Calibrated for precision measurements
- Resettable digital counter with two displays for individual upward and downward measurements