Learn More
MSC SPI Electronic Depth Gage, 0mm to 150mm, ABS Plastic, With Single Hook
Supplier: MSC 69547602
SPI Electronic Depth Gage is designed for precise measurements, offering a measuring range of 0mm to 150mm. This gage features a single hook for versatile measurements and includes both inch/metric conversion, absolute and incremental measuring modes for user convenience.
- Single hook for versatile measurements
- NIST traceability certification
- SPC/USB data output
- Automatic shut-off
- Linear scale embedded
- Inch/metric conversion
- Absolute and incremental measuring modes
- Short form calibration certificate included
- Furnished in a fitted case
- Two-year limited warranty