Learn More
MSC SPI Electronic Linear Scale, 0 to 12 in, 0.0015 in Accuracy
Supplier: MSC 35650316
This electronic linear scale provides precise horizontal and vertical measurements from 0 to 12 inches with 0.0015 inch accuracy and 0.0005 inch resolution. It has data output, IP54 protection, and an absolute encoder, eliminating zero setting upon power-up. Features include a large LCD, automatic on/off, and inch/metric/fraction conversion. Made of stainless steel, it includes an articulating arm, mounting brackets, and a fitted case.
- IP54 level protection
- Absolute encoder, no zero setting needed
- SPC/USB data output
- Large LCD display
- Articulating arm for multiple displays
- Mounting brackets included
- Stainless steel construction
- Automatic on/off for battery life
- Inch/metric/fraction conversion
- Furnished in fitted case
- Two year limited warranty
- D.O.T. regulated, ground freight only