Learn More
MSC SPI Gage Block Gage Block Accessory Set, For Use with Rectangular Gage Blocks, 14 Pieces
Supplier: MSC 50967108
This accessory set is designed to complement rectangular gage blocks, providing a versatile solution for precision measurement tasks. It includes 14 pieces, enhancing the functionality and application range of existing gage block sets.
- For use with rectangular gage blocks
- Includes an aluminum case for storage
- Features holders with capacities of 8.500 in L x 6.000 in and 4.175 in L x 2.250 in
- Comes with two pairs of beveled edge jaws (4 in and 6 in long)
- Includes four pairs of cylindrical jaws (0.100, 0.200, 0.250, and 0.500 in radius)
- Contains a total of 14 pieces
- Manufactured in China
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.