Learn More
MSC SPI Gage Block, Rectangle, Steel, 0.104 in, Grade 0
Supplier: MSC 86281250
This precision gage block is designed for accurate measurement in various industrial and laboratory applications. Made from durable steel, it features a rectangular shape and provides a high accuracy of Grade 0, ensuring reliable and consistent measurements.
- Rectangle shape
- Grade 0 accuracy
- Steel material
- Size: 0.104 in
- Includes NIST Certification