Learn More
MSC SPI Optical Comparator Chart & Reticle, 1-3/8 in Dia, 10x Magnification
Supplier:  MSC 06554059
Encompass
This optical comparator chart and reticle features a 1-3/8 inch diameter and is designed for use with 10x magnification. It is suitable for various measurement and inspection applications.
  • Designed for 10x magnification
  • Overall diameter of 1-3/8 in
  • Reticle number 5
  • Manufactured in Japan
Catalog No. 50-005-28450