Learn More
MSC SPI Pin Gage, 0.474 in Diameter, Class ZZ Minus, Black Oxide
Supplier:  MSC 56248230
Encompass
This is a Class ZZ Minus Plug & Pin Gage designed for precise measurement. It features a 0.474 inch diameter, a black oxide coating for corrosion prevention and wear indication, and is constructed from hardened drill rod with a Rockwell hardness of RC 60-62, ensuring durability and accuracy with a 0.000200 inch tolerance.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod RC 60-62.
  • 0.474 inch pin size.
  • 2 inch length.
  • 0.000200 inch tolerance.
Catalog No. 50-005-31827