Learn More
MSC SPI Pin Gage Minus, Black Oxide, 0.908 Inch Diameter, Hardened Drill Rod
Supplier:  MSC 56252596
Encompass
This pin gage features a 0.908 in diameter and a black oxide coating, which helps prevent corrosion and indicates gage wear. It is constructed from hardened drill rod with an RC 60-62 hardness, ensuring durability and accuracy for various measurement applications.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod RC 60-62.
  • 0.908 in diameter pin.
  • 2 in length.
Catalog No. 50-005-31843