Learn More
MSC SPI Plug Gage, 0.4710 in, Class ZZ Plus, Black Oxide
Supplier:  MSC 84503119
Encompass
This plug gage is designed for precise measurement applications. It features a 0.4710 inch diameter and a Class ZZ Plus tolerance, ensuring high accuracy for quality control and inspection tasks. The gage is finished with black oxide for enhanced durability and corrosion resistance.
  • Provides highly accurate dimensional checks.
  • Features a 0.4710 inch diameter for specific measurements.
  • Includes a Class ZZ Plus tolerance for precision.
  • Enhanced durability with a black oxide finish.
Catalog No. 50-005-27351