Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0620 in Diameter, Steel
Supplier: MSC 78382470
The Class ZZ Plus Plug Gage is engineered for precision measurement and inspection. It features a 0.0620 inch diameter with a tight tolerance of 0.000200 inch, making it suitable for demanding laboratory and industrial applications. Constructed from durable steel, this gage ensures lasting performance and consistent quality.
- Features a tight tolerance of 0.000200 inch for accurate measurements
- Made from steel for lasting performance
- Manufactured to Class ZZ Plus standards