Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2120 Dia, 0.000200 Tolerance
Supplier: MSC 78383973
This SPI Class ZZ Plus Plug Gage is designed for precise measurement applications, featuring a diameter of 0.2120 inches and a tolerance of 0.000200 inches. It is suitable for accurately checking hole sizes and depths in various industrial and manufacturing settings.
- Precision plug gage for accurate measurements
- Class ZZ Plus for stringent tolerance requirements
- Diameter of 0.2120 inches
- Tolerance of 0.000200 inches
- Ideal for quality control and inspection tasks