Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2160 in Diameter, 0.0002 in Tolerance
Supplier: MSC 78384013
This product is a Class ZZ Plus plug gage designed for precise measurement applications. It features a 0.2160 inch diameter and a tight tolerance of 0.000200 inch, constructed from durable steel.
- Precision measurement with a 0.2160 in diameter.
- Accurate tolerance of 0.000200 in.
- Durable steel construction for longevity.