Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.3330 in, Black Oxide
Supplier:  MSC 50742055
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a diameter of 0.3330 in and a tolerance of 0.000200 in. It comes with a black oxide coating/finish, ensuring durability for various inspection and calibration tasks.
  • Designed for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a black oxide coating/finish.
  • Made in China.
Catalog No. 50-005-28154