Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.4360 in, 0.000200 in Tolerance
Supplier:  MSC 78386216
Encompass
This Class ZZ Plus plug gage features a 0.4360 in diameter and a tolerance of 0.000200 in. Constructed from steel, it is designed for precise measurement applications.
  • Features a 0.4360 in diameter of measurement.
  • Offers a precise tolerance of 0.000200 in.
  • Constructed from durable steel.
  • Manufactured in China.
Catalog No. 50-005-27272