Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SPI Plug Gage, Class ZZ Plus, 0.4390 in Dia, 0.000200 in Tolerance

Supplier:  MSC 78386240

Encompass

This Class ZZ Plus Plug Gage is engineered for high-precision measurement tasks. It features a specific diameter and tight tolerance, providing a reliable solution for verifying hole sizes and ensuring accuracy in manufacturing and quality control processes.

  • Precise diameter of 0.4390 inch
  • Tight tolerance of 0.000200 inch
  • Constructed from durable steel
  • Country of origin: China

Catalog No. 50-005-30498


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.