Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.2550 in. Diameter, 0.000200 in. Tolerance, 1 ea
Supplier:  MSC 78384401
Encompass
This Class ZZ Plus plug gage is a precision measurement tool designed for checking hole sizes and other internal dimensions. Constructed from high-quality steel, it provides reliable performance in industrial inspection and quality control applications.
  • Diameter of 0.2550 in.
  • Tolerance of 0.000200 in.
  • Constructed from durable steel
Catalog No. 50-005-11739