Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.0760 in. Diameter, 1 ea
Supplier: MSC 78372729
The class ZZ minus plug and pin gage is a precision inspection tool designed for accurate diameter measurements and go/no-go gauging applications. Engineered from durable steel, this gage provides reliable performance in metalworking and industrial inspection environments.
- Diameter of measurement: 0.0760 in.
- Tolerance: 0.000200 in.
- Construction material: Steel