Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.2910 in. Diameter, 1 ea.
Supplier: MSC 78384765
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. It is constructed from durable steel to ensure reliable performance during go/no-go gauging tasks.
- Constructed from steel for durability
- Diameter of 0.2910 in.
- Tolerance of 0.000200 in.