Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.6050 in. Diameter, 0.0002 in. Tolerance, 1 ea
Supplier: MSC 78387909
This Class ZZ Plus plug gage is designed for precise hole measurement and inspection applications. Constructed from durable steel, this gage provides a reliable standard for checking dimensions in metalworking and manufacturing environments.
- Gauge diameter of 0.6050 in.
- Tolerance of 0.0002 in.
- Material composition of steel
- Suitable for go/no go inspection tasks