Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SPI Plug Gage, Class ZZ Plus, Steel, 0.9450 in. Diameter, 1 ea

Supplier:  MSC 78391307

Encompass

This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Constructed from durable steel, it ensures reliability and accuracy for dimensional verification requirements.

  • Features a 0.9450 in. diameter
  • Provides a 0.0002 in. tolerance
  • Manufactured from high-quality steel
  • Suitable for industrial quality control environments

Catalog No. 50-005-10811


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.