Learn More
MSC SPI Swiss Precision Instruments Class ZZ Plus Plug Gage, 0.147 in. Dia, 0.0002 in. Tolerance, 1 ea.
Supplier:  MSC 78383320
Encompass
This Class ZZ plus plug gage is a high-precision measurement tool designed for accurate hole diameter verification. Constructed from durable steel, this gage provides reliable performance in industrial metrology applications.
  • Diameter of 0.1470 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
  • Manufactured in the USA
Catalog No. 50-005-16107