Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.739 in. Diameter, 0.0002 in. Tolerance, 1 ea.
Supplier:  MSC 78389244
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and verification of hole diameters. Engineered for durability, it is constructed from high-quality steel to ensure consistent performance in various gaging applications.
  • Diameter of 0.7390 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
  • Made in the USA
Catalog No. 50-005-15997