Learn More
MSC SPI Plug and Pin Gage, Steel, 0.6280 in. Diameter, Class ZZ Minus, 1 ea.
Supplier:  MSC 78378247
Encompass
This Class ZZ plug gage is designed for precise measurement applications, featuring a diameter of 0.6280 in. and constructed from durable steel for reliable performance in industrial inspection environments.
  • Class ZZ accuracy
  • Minus tolerance of 0.0002 in.
  • Constructed from steel
  • Designed for go/no go inspection
Catalog No. 50-005-15170