Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.5940 in. Diameter, 1 ea.
Supplier:  MSC 78387792
Encompass
This Class ZZ Plus plug gage is a high-precision measurement tool designed for accurate quality control and inspection applications in industrial metalworking environments. Constructed from durable steel, this gage is engineered to provide reliable performance when checking hole sizes.
  • Diameter of 0.5940 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel for durability
Catalog No. 50-005-11745