Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0330 in Diameter, Black Oxide
Supplier: MSC 56243801
This plug and pin gage is a Class ZZ Minus gage with a 0.0330 inch diameter. It is designed for precise measurement applications.
- Class ZZ minus
- Black oxide coating/finish
- Used for setting micrometers, NoGo inspection, and other measuring devices
- Includes certificate of accuracy with NIST traceability