Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4350 in Diameter, Black Oxide
Supplier: MSC 56247844
This Class ZZ Minus plug and pin gage is designed for precise measurement applications. It features a 0.4350 in diameter and a black oxide coating, making it suitable for various industrial and laboratory settings.
- Provides accurate measurements with a 0.4350 in diameter.
- Features a Class ZZ Minus tolerance for high precision.
- Durable black oxide finish for extended tool life.
- Ideal for quality control and inspection processes.