Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.8370" Dia, Black Oxide
Supplier:  MSC 56251879
Encompass
This plug and pin gage is a Class ZZ Minus precision tool, featuring a 0.8370 inch diameter and a durable black oxide finish.
  • Diameter of 0.8370 inch
  • Tolerance of 0.000200 inch
  • Black oxide coating/finish
  • Manufactured in China
Catalog No. 50-005-31805