Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.9730 Dia, Black Oxide
Supplier: MSC 85666303
This plug and pin gage is designed for precise measurement applications. It features a 0.9730 inch diameter, a Class ZZ minus tolerance, and a durable black oxide finish. This gage is ideal for verifying hole sizes and depths with high accuracy.
- Class ZZ minus tolerance for critical measurements
- 0.9730 inch diameter ensures accuracy
- Black oxide coating for enhanced durability and corrosion resistance
- Tolerance of 0.000200 inch for high precision