Learn More
MSC SPI Plug Thread Gage, 5/16-32 Thread, 3B Class, Double End, Go & No Go
Supplier: MSC 75889360
This SPI Plug Thread Gage features a 5/16-32 thread size and 3B class for precise measurements. It is a double-ended gage with both "Go" and "No Go" functions, ensuring comprehensive thread inspection. Constructed from durable steel, the gage includes a handle for ease of use.
- Go and No Go functions
- 5/16-32 thread size
- 3B classification
- Double end design
- Durable steel construction
- Handle included
- NIST traceability certification
- Adheres to NIST standards
- Manufactured in India