Learn More
MSC SPI Plug Thread Gage, #6-32 Thread, 3B Class, No Go, Taperlock
Supplier:  MSC 55313571
Encompass
This SPI Plug Thread Gage is designed for the precise inspection of internal threads, ensuring proper assembly of mating components. It features a #6-32 thread, 3B class, and a No Go designation, making it suitable for checking the upper limit of the thread tolerance. Constructed from durable tool steel, this taperlock gage provides reliable and accurate measurements.
  • Certificate traceable to NIST standards
  • Thread measurements as per ANSI specification 1.16B
  • Through hardened RC 60-64
  • Handles not included with single members
  • Made from tool steel
  • Designed for internal thread inspection
  • No Go functionality for checking thread tolerance
Catalog No. 50-005-28620