Learn More
MSC SPI Plug Thread Gage, 7/8-14 Thread, 3B Class, Single End, No Go
Supplier:  MSC 55309298
Encompass
This plug thread gage is designed for inspecting internal threads. It features a 7/8-14 thread with a 3B class and is a single-end, No Go type gage. Constructed from tool steel, it has a right-hand thread direction. An inspection certificate traceable to NIST standards (ASME B1.2) is included for this non-calibrated gage.
  • Taperlock design
  • Inspection of internal threads to ensure assembly of mating components
  • Made from durable tool steel
  • Includes an inspection certificate traceable to NIST standards (ASME B1.2)
  • Single-end, No Go type
  • Right-hand thread direction
Catalog No. 50-005-28267