Learn More
MSC SPI Plug Thread Gage, M14x2 Thread, 6H Class, Double End, Go & No Go
Supplier:  MSC 54334719
Encompass
This plug thread gage is designed for the inspection of internal threads, ensuring the correct assembly of mating components. It features an M14x2 thread with a 6H class, offering both 'Go' and 'No Go' measurements in a double-ended configuration. A handle is included for ease of use.
  • Taperlock design
  • Facilitates inspection of internal threads
  • Ensures proper assembly of mating components
  • Supports Go and No Go measurements
  • M14x2 thread size
  • Constructed from tool steel
  • 6H classification
  • Right hand thread direction
  • Double-ended design
  • Metric thread standard
  • Includes handle
Catalog No. 50-005-28573