Learn More
MSC SPI Plug Thread Gage, M16x1.5, 6H Class, Double End
Supplier:  MSC 54334685
Encompass
This plug thread gage is designed for the inspection of internal M16x1.5 threads with a 6H class to ensure the proper assembly of mating components. It features a double-end design and is constructed from durable tool steel, with a handle included for ease of use.
  • Double-end design for comprehensive inspection
  • Inspects M16x1.5 threads with 6H class
  • Features both Go and No Go ends
  • Constructed from tool steel for durability
  • Includes handle for easy operation
  • Right hand thread direction
  • Supplied with an inspection certificate traceable to NIST standards
Catalog No. 50-005-27771