Learn More
MSC SPI Plug Thread Gage, M18x2.5 Thread, 6H Class, Double End
Supplier: MSC 54334677
This plug thread gage is designed for the inspection of internal threads, ensuring the proper assembly of mating components. It features a M18x2.5 thread size and 6H class, made from durable tool steel.
- Handle included
- Taperlock design
- Double end configuration
- Go and No Go functionality
- Inspection of internal threads to ensure assembly of mating components
- Metric thread standard